Learn More
Sensidyne™ Detector Tubes: Water Vapor
Supplier: Sensidyne™ 177UL
Description
All detector tubes are sold in packs of 10.
Specifications
Water Vapor | |
10 | |
32°F to 104°F |
3 to 80 lb./MMcf | |
1 min. | |
0°C to 40°C |