Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate

Catalog No. STGN4F
Click to view available options
Quantity:
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Perform accurate AST of non-fastidious Gram-negative isolates with this convenient plate

Confidently perform susceptibility testing of non-fastidious Gram-negative isolates with Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate.

Confidently perform susceptibility testing of non-fastidious Gram-negative isolates with Thermo Scientific™ Sensititre™ Gram Negative GN4F AST Plate.

  • Automated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
  • Individual packaging allows the laboratory to test one plate at a time with no waste

Not all products are available for sale in all territories. Please inquire.
 

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
AST Type MIC
Broth Type MHB (Part No. T3462)
Concentration Multiple
Description Sensititre Gram Negative MIC Plate, with Doripenem and Tigecycline
Antibiotic Code Various
Format Multi-antibiotic Microtitre Panels
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type 10 x Microtitre Panels
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.