Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Life Sciences™ Sentino Membrane Dispenser Refill ME25

Catalog No. 09927590 Shop All Cytiva Products
Click to view available options
Pore Size:
0.45 μm
This item is not returnable. View return policy

Catalog No. 09-927-590

Supplier: Cytiva Life Sciences™ 68121ME

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gridded, ME25 mixed cellulose ester membranes.

Sentino Membrane Dispenser Refill ME25

Sentino Membrane Dispenser Refill ME25
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Sentino Filter Dispenser
Material MCE
Pore Size 0.45 μm
Sterility Gamma-irradiated
Membrane Type Gridded
Model 68121ME
Type Membranes, Sentino Dispenser Pack
Diameter (Metric) 47 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.