Learn More
Kimberly-Clark™ Professional Sentry III 6pt Ratchet Hard Hat
Catalog No. p-4706497
Encompass
Change view
Click to view available options
Size:
6.5 to 8
Color:
Green
Orange
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
17-350-1F 6.5 to 8 Orange
19-804-230 6.5 to 8 Red
17-350-1D 6.5 to 8 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-350-1F Supplier Kimberly-Clark™ Professional Supplier No. 14420
Only null left
Add to Cart
Add to Cart
Kimberly-Clark™ Professional Sentry III 6pt Ratchet Hard Hat are intended to reduce the force of impact that could result from a blow to the top of the head
  • Low-profile, silhouette-styled
  • Meet ANSI Z89.1 - 2009 requirements for impact (as well as Class C, G and E)
  • Six-point ratchet suspension system and a flat, customizable front area
  • Customize to suit needs: Fits head sizes 6.5 to 8 and have accessory slots for attaching additional compatible personal protective equipment
Specifications
Color Orange
Size 6.5 to 8