Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569531 Supplier Invitrogen™ Supplier No. PA569531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%; Zebrafish: 100%.

The protein encoded by this gene belongs to the SEP15/selenoprotein M family. The exact function of this protein is not known; however, it has been found to associate with UDP-glucose:glycoprotein glucosyltransferase (UGTR), an endoplasmic reticulum(ER)-resident protein, which is involved in the quality control of protein folding. The association with UGTR retains this protein in the ER, where it may play a role in protein folding. It has also been suggested to have a role in cancer etiology. This protein is a selenoprotein, containing the rare amino acid selenocysteine (Sec). Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEP15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Selenof
Gene Accession No. Q923V8
Gene Alias 15 kDa selenoprotein; 15 kDa selenoprotein isoform 1 precursor; 15 kDa selenoprotein SeP15; 15kDa selenoprotein; 15-kDa selenoprotein; 15-Sep; 9430015P09Rik; cb29; Selenof; selenoprotein; selenoprotein 15; selenoprotein F; Sep15; sep15 protein; si:dkey-43b14.10; zgc:86882
Gene Symbols Selenof
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat SEP15.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113922
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.