Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103009
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103009 Supplier Invitrogen™ Supplier No. PA5103009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SEP15.

The protein encoded by this gene belongs to the SEP15/selenoprotein M family. The exact function of this protein is not known; however, it has been found to associate with UDP-glucose:glycoprotein glucosyltransferase (UGTR), an endoplasmic reticulum(ER)-resident protein, which is involved in the quality control of protein folding. The association with UGTR retains this protein in the ER, where it may play a role in protein folding. It has also been suggested to have a role in cancer etiology. This protein is a selenoprotein, containing the rare amino acid selenocysteine (Sec). Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEP15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Selenof
Gene Accession No. O60613, Q923V8
Gene Alias 15 kDa selenoprotein; 15 kDa selenoprotein isoform 1 precursor; 15 kDa selenoprotein SeP15; 15kDa selenoprotein; 15-kDa selenoprotein; 15-Sep; 9430015P09Rik; cb29; Selenof; selenoprotein; selenoprotein 15; selenoprotein F; Sep15; sep15 protein; si:dkey-43b14.10; zgc:86882
Gene Symbols Selenof
Host Species Rabbit
Immunogen A synthesized peptide derived from human SELENOF(Accession O60613), corresponding to amino acid residues E35-L63.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113922, 9403
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.