Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Separase Antibody (XJ11-1B12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100439MFV500
Description
Separase Monoclonal antibody specifically detects Separase in Human samples. It is validated for Western BlotSpecifications
Separase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
XJ11-1B12 | |
Western Blot | |
Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase | |
Maltose-Binding Protein fusion of a C-terminal fragment of human Separase (residues 1866-1996). [UniProt# Q14674] | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, Mitotic Regulators, Phospho Specific | |
9700 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction