Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Separase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987967
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7967 Supplier Bioss Supplier No. BS7732R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Caspase-like protease, which plays a central role in the chromosome segregation by cleaving the SCC1/RAD21 subunit of the cohesin complex at the onset of anaphase. During most of the cell cycle, it is inactivated by different mechanisms.

Specifications

Antigen Separase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ESPL1
Gene Accession No. Q14674
Gene Alias AL024103; AU045071; Caspase-like protein ESPL1; Cerp; Cut1/ESP1 related protein; ESP1; ESPL1; extra spindle pole bodies 1 (S. cerevisiae); extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle pole bodies like 1, separase; extra spindle poles like 1; Extra spindle poles-like 1 protein; FLJ46492; Kiaa0165; PRCE; SEPA; Separase; Separin; separin, cysteine protease; SSE
Gene Symbols ESPL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Separase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9700
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.