Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sephacryl™ High Resolution Media

Catalog No. 45002057 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
150 mL
750 mL
Product Type:
Sephacryl S-100 HR media
Sephacryl S-200 HR media
Sephacryl S-300 HR media
Sephacryl S-400 HR media
Flow Rate:
>125 cm/hr.
>150 cm/hr.
Particle Size:
25 to 75μm
50μm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Flow Rate Particle Size
45-002-057 150 mL Sephacryl S-300 HR media >150 cm/hr. 50μm
45-002-061 750 mL Sephacryl S-100 HR media >125 cm/hr. 25 to 75μm
45-002-056 750 mL Sephacryl S-400 HR media >150 cm/hr. 50μm
45-002-059 150 mL Sephacryl S-200 HR media >150 cm/hr. 50μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45-002-057 Supplier Cytiva Supplier No. 17059910
Add to Cart
Add to Cart

Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.

  • Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
  • Gel filtration media providing high resolution with fast and easy operation
  • Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Long-term physical, chemical stability and easy handling
  • Hydrophilic nature enables maximum recovery and minimum     nonspecific adsorption

  • Average particle Size: 50μm
  • Matrix: Allyl dextran and N, N'-methylenebisacrylamide
  • pH stability working range: 3-11
  • Shelf Life: 5years

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place)
Product Type Sephacryl S-300 HR media
Quantity 150 mL
Height (English) 5.9 in.
Height (Metric) 15cm
Flow Rate >150 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 11
Particle Size 50μm
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Content And Storage 4° to 30°C
For Use With (Application) Purification of Proteins, Polysaccharides and other macromolecules
Matrix Gel Filtration Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.