Learn More
Description
Established medium for desalting and buffer exchange
- Gel filtration medium
- Available in coarse, medium, fine and superfine grades
- All grades satisfy process-scale chromatography requirements
- Available in lab packs or prepacked columns
Specifications:
- Working pH range: 2 to 13
- Bed volume (in distilled water): 4 to 6mL/g
- Fractionation range dextrans/globular proteins: 1× 102 to 5 × 103 / 1 × 103 to 5 × 103 Mr
- Max. flowrate/operating pressure: the beads behave as rigid spheres obeying Darcy's Law
Specifications
Specifications
| Quantity | 100 g |
| Flow Rate | >47 cm/hr. |
| Particle Size | 17 to 132 μm (Wet Form), >20 μm (Dry Form) |
| Bed Volume | 4 to 6mL/g in Distilled Water |
| Product Type | Sephadex G-25 Media |
| Format | Solid Spherical Bead |
| Diameter (Metric) Inner | 2.6cm |
| For Use With (Application) | Desalting and Buffer exchange and removal free label in industrial applications |
| Height (English) | 11.81 in. |
| Height (Metric) | 30cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.