Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Sephadex™ G-25, MP Biomedicals™

Used in affinity chromatography, protein chromatography and gel filtration chromatography. Sephadex™ G-25, MP Biomedicals is a gel filtration medium that has been used to study the acceleration of diabetic wound healing and the treatment of epitheli

$101.00 - $1865.00

Specifications

Storage Requirements Store at Room Temperature (15–30°C).
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
02-195-25410
View Documents Promotion Details
MP Biomedicals™
0219525410
10g
Each for $101.00
Add to cart
 
02-195-25450
View Documents Promotion Details
MP Biomedicals™
0219525450
50g
Each for $358.00
Add to cart
 
02-195-25480
View Documents Promotion Details
MP Biomedicals™
0219525480
100g
Each for $1,865.00
Add to cart
 
Description

Description

Specifications

Specifications

Store at Room Temperature (15–30°C).
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit