Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sephadex™ Ion Exchange Media

Catalog No. 45000042 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 g
500 g
Material:
CM-Sephadex C-25
CM-Sephadex C-50
DEAE-Sephadex A-25
DEAE-Sephadex A-50
QAE-Sephadex A-25
QAE-Sephadex A-50
SP-Sephadex C-25
MWCO:
30,000 to 100,000
Up to 30,000
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Material MWCO
45-000-042 500 g CM-Sephadex C-25 Up to 30,000
45-000-040 100 g QAE-Sephadex A-50 30,000 to 100,000
45-000-037 500 g DEAE-Sephadex A-50 30,000 to 100,000
45-000-034 100 g DEAE-Sephadex A-25 Up to 30,000
45-000-035 500 g DEAE-Sephadex A-25 Up to 30,000
45-000-036 100 g DEAE-Sephadex A-50 30,000 to 100,000
45-000-038 100 g QAE-Sephadex A-25 Up to 30,000
45-000-041 100 g CM-Sephadex C-25 Up to 30,000
45-000-044 500 g CM-Sephadex C-50 30,000 to 100,000
45-000-043 100 g CM-Sephadex C-50 30,000 to 100,000
45-000-045 100 g SP-Sephadex C-25 Up to 30,000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 45-000-042 Supplier Cytiva Supplier No. 17021002
Add to Cart
Add to Cart

Separate molecules over broad molecular weight and pH ranges

Range of weak and strong anionic (A) and cationic (C) exchangers. Fractionation range: A-25 and C-25, up to 30 000; A-50 and C-50: 30 000–100 000.

Very high capacities even at elevated ionic strengths
Very low non-specific adsorption
Wide range of applications at laboratory and process scales
Suitable for column or batch techniques
Simple and economical to use

DEAE Sephadex

  • Weak and strong anion exchanger
  • pH range 2-9

QAE Sephadex
  • Strong anion exchanger
  • pH range: 2-12
CM Sephadex
  • Weak cation exchanger
  • pH range: 6-10
SP Sephadex
  • Strong cation exchanger
  • pH range: 4-13

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 g
Particle Size 40 to 120 μm in dry form
For Use With (Equipment) Column or Batch Techniques
Product Type Sephadex Ion Exchange Media
Format Solid Spherical Bead
For Use With (Application) Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
Height (English) 2 to 5.9 in.
Height (Metric) 5 to 15cm
Ligand Type Carboxymethyl
Material CM-Sephadex C-25
pH Range 6 to 10
Support Type Cross-linked Dextran
MWCO Up to 30,000
Content And Storage 4° to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.