Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sepharose Agarose Gel Filtration Media

Catalog No. 45000031 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
<11.5 cm/hr.
<14 cm/hr.
<15 cm/hr.
<26 cm/hr.
<30 cm/hr.
Particle Size:
45 to 165 μm
60 to 200 μm
pH Range:
3 to 12
3 to 13
4 to 9
Support Type:
2% Agarose
4% Agarose
6% Agarose
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Particle Size pH Range Support Type
45-000-031 <15 cm/hr. 60 to 200 μm 3 to 13 2% Agarose
45-000-032 <26 cm/hr. 45 to 165 μm 3 to 13 2% Agarose
45-000-029 <11.5 cm/hr. 45 to 165 μm 4 to 9 4% Agarose
45-000-033 <30 cm/hr. 45 to 165 μm 3 to 12 6% Agarose
45-000-028 <14 cm/hr. 45 to 165 μm 4 to 9 6% Agarose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-000-031 Supplier Cytiva Supplier No. 17014001

Choose from a wide range of bead-formed agarose-based gel filtration media. Cytiva Life Sciences™ Sepharose Agarose Gel Filtration Media shows good separation over a wide molecular weight range.

European Pharmacopoeia Solutions

  • Reagents and Standard solutions in Chapters 2 and 4 of the current Ph. Eur.
  • All are manufactured and tested in compliance with the Ph. Eur.
  • Supplied with a Certificates of Analysis
  • Lot No. and expiry date are stated on all product labels

Order Info

Supplied in 1L plastic containers

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Flow Rate <15 cm/hr.
Particle Size 60 to 200 μm
Product Type Sepharose Agarose Gel Filtration Media
Format Solid Bead
Diameter (Metric) Inner 5cm
For Use With (Application) Allow characterization or cleaning up of samples containing components of diverse molecular weight
Height (English) 3.94 in.
Height (Metric) 10cm
Max. Pressure 60-120 cm/h, pressure drop cm H2O/bed height=15, bed height 10 cm, 5 cm i.d.
pH Range 3 to 13
Support Type 2% Agarose
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.