Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPHS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116562 Supplier Invitrogen™ Supplier No. PA5116562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: BT-474, SW620, HepG2, U-251MG, Mouse liver, Mouse kidney, Rat liver, Rat kidney Immunogen sequence: MSTRESFNPE SYELDKSFRL TRFTELKGTG CKVPQDVLQK LLESLQENHF QEDEQFLGAV MPRLGIGMDT CVIPLRHGGL SLVQTTDYIY PIVDDPYMMG RIACANVLSD LYAMGVTECD NMLMLLGVSN KMTDRERDKV MPLIIQGFKD AAEEAGTSVT GGQTVLNPWI VLGGVATTVC QPNEFIMPDN AVPGDVLVLT KPLGTQVAVA VHQWLDIPEK WNKIKLVVTQ EDVELAYQEA MMNMARLNRT AAGLMHTFNA HAATDITGFG ILGHAQNLAK QQRNEVSFVI HNLPVLAKMA AVSKACGNMF GLMHGTCPET SGGLLICLPR EQAARFCAEI KSPKYGEGHQ AWIIGIVEKG NRTARIIDKP RIIEVAPQVA TQNVNPTPGA TS.

This protein encodes an enzyme that synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPHS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SEPHS1
Gene Accession No. P49903, Q8BH69
Gene Alias 1110046B24Rik; AA589574; AI505014; AW111620; RP11-24J20.2; SELD; selenide; Selenide, water dikinase 1; selenium donor protein 1; Selenophosphate synthase 1; selenophosphate synthetase 1; selenophosphate synthetase 1 +E9; selenophosphate synthetase 1 +E9a; selenophosphate synthetase 1 delta E2; selenophosphate synthetase 1 delta E8; selenophosphate synthetase 1 major; SEPHS1; SPS; SPS1
Gene Symbols SEPHS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-392 of human SEPHS1 (NP_0363792).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109079, 22929, 291314
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.