Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPHS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527950 Supplier Invitrogen™ Supplier No. PA527950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (87%), Dog (96%), Pig (95%), Rabbit (96%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (95%), Guinea pig (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This protein encodes an enzyme that synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons. This protein itself contains a selenocysteine residue in its predicted active site. The 3' UTR of the gene has a stem-loop secondary structure called a selenocysteine insertion sequence (SECIS) element, which allows UGA to direct the incorporation of selenocysteine rather than signal a translational stop.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPHS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SEPHS2
Gene Accession No. Q99611
Gene Alias clone 1000; selenide; selenide, water dikinase 2; Selenium donor protein 2; selenophosphate synthase 2; selenophosphate synthetase 2; Sephs2; Sps2; SPS2b; the SelD gene product; Ysg3
Gene Symbols SEPHS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SEPHS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.