Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPHS2 Polyclonal Antibody
SEPHS2 Polyclonal Antibody
Supplier: Thermo Scientific PA583888
Description
SEPHS2 Polyclonal Antibody for ICC/IF, IHC (P)

Specifications
SEPHS2 | |
Polyclonal | |
Unconjugated | |
SEPHS2 | |
clone 1000; selenide; selenide, water dikinase 2; Selenium donor protein 2; selenophosphate synthase 2; selenophosphate synthetase 2; Sephs2; Sps2; SPS2b; the SelD gene product; Ysg3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22928 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99611 | |
SEPHS2 | |
Recombinant protein corresponding to Human SEPHS2. Recombinant protein control fragment (Product #RP-100770). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction