Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPINEO (TM) P 600, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical SEPINEO (TM) P 600
Specifications
Specifications
Color | Opaque, White, Yellow tint |
pH | 5 to 7 |
Physical Form | Liquid |
Quantity | 500 mL |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction