Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEPN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543082
Description
Peptide sequence: ANYFLDITSV KPEEIESNLF SFSSTFEDPS TATYMQFLKE GLRRGLPLLQ Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rat: 100%; Zebrafish: 79%.
SEPN1 is a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. Mutations in SEPN1 gene cause the classical phenotype of multiminicore disease and congenital muscular dystrophy with spinal rigidity and restrictive respiratory syndrome. This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Mutations in this gene cause the classical phenotype of multiminicore disease and congenital muscular dystrophy with spinal rigidity and restrictive respiratory syndrome. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| SEPN1 | |
| Polyclonal | |
| Unconjugated | |
| SELENON | |
| 1110019I12Rik; AI414492; cb686; CFTD; FLJ24021; MDRS1; RSMD1; RSS; selenon; Selenoprotein N; selenoprotein N, 1; SELN; SePN; sepn1; wu:fb06g01; wu:fb73d02; wu:fv41b08; zgc:101091 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57190 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NZV5 | |
| SELENON | |
| Synthetic peptide directed towards the C-terminal of human SEPN1 (aa 540-589). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction