Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563520 Supplier Invitrogen™ Supplier No. PA563520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WSLVKELEEL QNKQENSSHQ KLAGLHLEKY SFPVEMMICL PNGTVVHHIN ANYFLDITSV KPEEIESNLF SFSSTFEDPS T Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

SEPN1 is a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. Mutations in SEPN1 gene cause the classical phenotype of multiminicore disease and congenital muscular dystrophy with spinal rigidity and restrictive respiratory syndrome. This gene encodes a selenoprotein, which contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon that normally signals translation termination. The 3' UTR of selenoprotein genes have a common stem-loop structure, the sec insertion sequence (SECIS), that is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Mutations in this gene cause the classical phenotype of multiminicore disease and congenital muscular dystrophy with spinal rigidity and restrictive respiratory syndrome. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SELENON
Gene Accession No. Q9NZV5
Gene Alias 1110019I12Rik; AI414492; cb686; CFTD; FLJ24021; MDRS1; RSMD1; RSS; selenon; Selenoprotein N; selenoprotein N, 1; SELN; SePN; sepn1; wu:fb06g01; wu:fb73d02; wu:fv41b08; zgc:101091
Gene Symbols SELENON
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPN1. Recombinant protein control fragment (Product #RP-99821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.