Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557596 Supplier Invitrogen™ Supplier No. PA557596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHIPVYQQEE NQTDVWTLLN GSKDDFLIYD RCGRLVYHLG LPFSFLTFPY VEEAIKIAYC EKKCGNCSLT TLKD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

A growing body of evidence relates selenium to cancer prevention, immune system function, male fertility, cardiovascular disorder, control of the aging and neurodiseases process. Selenoproteins are thought to be responsible for the majority of these biomedical effects of selenium. Approximately 17 selenoproteins have been identified until now. Although the function of many selenoproteins are unknown, some play important roles in anti-oxidant mechanisms. It has been also implicated in the regulation of signaling pathways through catalysis of thiol/disulfide exchange. Selenoprotein M (SelM) is especially expressed in a mammalian brain and is localized to the perinuclear structures (Golgi/ER). The roles of selM have not been clearly identified until present time.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SELENOP
Gene Accession No. P49908
Gene Alias AU018766; D15Ucla1; Plasma selenoprotein P; Selenop; Selenoprotein P; selenoprotein P, plasma, 1; Selenoprotein Se-P1; Selenoprotein Se-P10; Selenoprotein Se-P2; Selenoprotein Se-P6; SELP; seP; Se-P; SEPP; SEPP1
Gene Symbols SELENOP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPP1. Recombinant protein control fragment (Product #RP-96987).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6414
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.