Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112707
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5112707 100 μg
1 options

Catalog No. PIPA5112707

Supplier: Invitrogen™ PA5112707

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEPP1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

A growing body of evidence relates selenium to cancer prevention, immune system function, male fertility, cardiovascular disorder, control of the aging and neurodiseases process. Selenoproteins are thought to be responsible for the majority of these biomedical effects of selenium. Approximately 17 selenoproteins have been identified until now. Although the function of many selenoproteins are unknown, some play important roles in anti-oxidant mechanisms. It has been also implicated in the regulation of signaling pathways through catalysis of thiol/disulfide exchange. Selenoprotein M (SelM) is especially expressed in a mammalian brain and is localized to the perinuclear structures (Golgi/ER). The roles of selM have not been clearly identified until present time.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SELENOP
Gene Accession No. P49908, P70274
Gene Alias AU018766; D15Ucla1; Plasma selenoprotein P; Selenop; Selenoprotein P; selenoprotein P, plasma, 1; Selenoprotein Se-P1; Selenoprotein Se-P10; Selenoprotein Se-P2; Selenoprotein Se-P6; SELP; seP; Se-P; SEPP; SEPP1
Gene Symbols SELENOP
Host Species Rabbit
Immunogen Recombinant Human Selenoprotein P protein (20-381AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20363, 6414
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.