Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 113701AP150UL

Description
Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintece of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer′s disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.Specifications
SEPT1 | |
Polyclonal | |
Unconjugated | |
SEPT1 | |
DIFF6, LARP, Peanut like protein 3, PNUTL3, SEP1, SEPT1, septin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1731, 293507, 54204 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42209, Q5EB96, Q8WYJ6 | |
SEPT1 | |
SEPT1 Fusion Protein Ag1926 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction