Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114098 Supplier Invitrogen™ Supplier No. PA5114098
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVAVGRPSN EELRNLSLSG HVGFDSLPDQ LVNKSTSQGF CFNILCVGET For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPT11
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SEPT11
Gene Accession No. Q9NVA2
Gene Alias 6230410I01Rik; AW548875; D5Ertd606e; SEPT11; Sept6; septin 11; septin 6; Septin11; septin-11
Gene Symbols SEPT11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SEPT11(septin 11) (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55752
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.