Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEPT12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598593
Description
SEPT12 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).
Specifications
SEPT12 | |
Polyclonal | |
Unconjugated | |
SEPT12 | |
1700028G04Rik; 4933413B09Rik; SEPT12; septin 12; SEPTIN12; septin-12; SPGF10; testicular tissue protein Li 168 | |
Rabbit | |
Protein G | |
RUO | |
124404, 71089 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8IYM1 | |
SEPT12 | |
Recombinant Human Septin-12 protein (1-358AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction