Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPT12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111132 Supplier Invitrogen™ Supplier No. PA5111132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHYENYRVIR LNESHLLPRG PGWVNLAPAS PGQLTTPRTF KVCRGAHDDS DDEF Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 26%.

Septins, such as SEPT12, are conserved GTP-binding proteins that function as dynamic, regulatable scaffolds for the recruitment of other proteins. They are involved in membrane dynamics, vesicle trafficking, apoptosis, and cytoskeleton remodeling, as well as infection, neurodegeneration, and neoplasia (Hall et al., 2005 [PubMed 15915442]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPT12
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPT12
Gene Accession No. Q8IYM1
Gene Alias 1700028G04Rik; 4933413B09Rik; SEPT12; septin 12; SEPTIN12; septin-12; SPGF10; testicular tissue protein Li 168
Gene Symbols SEPT12
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SEPT12. Recombinant protein control fragment (Product #RP-98649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124404
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.