Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPT2 Recombinant Rabbit Monoclonal Antibody (JB41-30), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA534651
Description
Positive Control: Mouse brain, K562, SHSY-5Y, HUVEC, HepG2, human colon cancer tissue, human kidney tissue, rat brain tissue.
Septin 2 is a filament-forming cytoskeletal GTPase. Septin 2 is required for normal organization of the actin cytoskeleton and plays a role in the biogenesis of polarized columnar-shaped epithelium by maintaining polyglutamylated microtubules, thus facilitating efficient vesicle transport, and by impeding MAP4 binding to tubulin. It is required for the progression through mitosis, and it forms a scaffold at the midplane of the mitotic splindle required to maintain CENPE localization at kinetochores and consequently chromosome congression. During anaphase, Septin 2 may be required for chromosome segregation and spindle elongation. Septin 2 also plays a role in ciliogenesis and collective cell movements.
Specifications
SEPT2 | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q15019 | |
SEPTIN2 | |
Recombinant protein within Human Septin 2 aa 182-318 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
JB41-30 | |
Unconjugated | |
SEPTIN2 | |
2-Sep; AW208991; DIFF6; hNedd5; KIAA0158; mKIAA0158; NEDD5; Nedd-5; Neural precursor cell expressed developmentally down-regulated protein 5; neural precursor cell expressed, developmentally down-regulated 5; neural precursor cell expressed, developmentally down-regulated gene 5; Pnutl3; Sept2; septin 2; Septin2; septin-2; Vascular endothelial cell specific protein 11; Vesp11 | |
Rabbit | |
Protein A | |
RUO | |
4735 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction