Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584621 Supplier Invitrogen™ Supplier No. PA584621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPLIAKADCL VPSEIRKLKE RIREEIDKFG IHVYQFPE Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. Disruption of septin function disturbs cytokinesis and results in large multinucleate or polyploid cells. This gene is mapped to 22q11, the region frequently deleted in DiGeorge and velocardiofacial syndromes. A translocation involving the MLL gene and this gene has also been reported in patients with acute myeloid leukemia. Two transcripts of this gene, a major one of 2.2 kb and a minor one of 3.5 kb, have been observed. The 2.2 kb form results from the utilization of a non-consensus polyA signal (AACAAT). In the absence of polyadenylation from this imperfect site, the consensus polyA signal of the downstream neighboring gene (GP1BB; platelet glycoprotein Ib) is used, resulting in the 3.5 kb transcript. An alternatively spliced transcript variant with a different 5' end has also been identified, but its full-length nature has not been completely determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPT5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPTIN5
Gene Accession No. Q99719
Gene Alias 5-Sep; CDCREL; CDCREL1; CDCREL-1; CDCrel-1A; cell division control related protein 1; cell division control-related protein 1; H5; HCDCREL-1; peanut-like 1; peanut-like 1 homolog; peanut-like protein 1; platelet glycoprotein Ib beta chain; Pnutl1; Sept5; septin 5; SEPTIN5; septin-5
Gene Symbols SEPTIN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPT5. Recombinant protein control fragment (Product #RP-103688).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5413
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.