Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SEPT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552177 Supplier Invitrogen™ Supplier No. PA552177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGKSTLMDTL FNTKFEGEPA THTQPGVQLQ SNTYDLQESN VRLKLTIVST VGFGDQINKE DSYKPIVEFI DAQFEAYLQE ELKIRRVLHT YHDSRIHVCL YFIAPTGHSL KSLDLVTMKK LDSKVNIIPI IAKADAISKS EL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

SEPT6 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. One version of pediatric acute myeloid leukemia is the result of a reciprocal translocation between chromosomes 11 and X, with the breakpoint associated with the genes encoding the mixed-lineage leukemia and septin 2 proteins. This gene encodes four transcript variants encoding three distinct isoforms. An additional transcript variant has been identified, but its biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SEPT6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPTIN6
Gene Accession No. Q14141
Gene Alias 2810035H17Rik; 2-Sep; 6-Sep; C920001C06Rik; hypothetical protein LOC540783; Kiaa0128; MGC16619; MGC20339; mKIAA0128; RP5-876A24.2; SEP2; Sep6; SEPT11; SEPT2; SEPT6; septin 11; septin 2; septin 6; SEPTIN6; septin-6
Gene Symbols SEPTIN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPT6. Recombinant protein control fragment (Product #RP-92139).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23157
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.