Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SEPT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106731
Description
Antibody detects endogenous levels of total SEPT6.
SEPT6 is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. One version of pediatric acute myeloid leukemia is the result of a reciprocal translocation between chromosomes 11 and X, with the breakpoint associated with the genes encoding the mixed-lineage leukemia and septin 2 proteins. This gene encodes four transcript variants encoding three distinct isoforms. An additional transcript variant has been identified, but its biological validity has not been determined.
Specifications
| SEPT6 | |
| Polyclonal | |
| Unconjugated | |
| SEPTIN6 | |
| 2810035H17Rik; 2-Sep; 6-Sep; C920001C06Rik; hypothetical protein LOC540783; Kiaa0128; MGC16619; MGC20339; mKIAA0128; RP5-876A24.2; SEP2; Sep6; SEPT11; SEPT2; SEPT6; septin 11; septin 2; septin 6; SEPTIN6; septin-6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23157, 56526, 691335 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14141, Q9R1T4 | |
| SEPTIN6 | |
| A synthesized peptide derived from human SEPT6(Accession Q14141), corresponding to amino acid residues V222-V272. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction