Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEPT8 Mouse anti-Human, Clone: 4A12G9, Proteintech
SDP

Catalog No. 501736622 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6622 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6622 Supplier Proteintech Group Inc Supplier No. 661881IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kDa GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 ).

Specifications

Antigen SEPT8
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4A12G9
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SEPT8
Gene Accession No. Q92599
Gene Alias KIAA0202, SEP2, SEPT8, septin 8
Gene Symbols SEPT8
Host Species Mouse
Immunogen SEPT8 Fusion Protein Ag19114
Purification Method Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23176
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.