Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-1 Antibody (OTI1E1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274072G
Description
Septin-1 Monoclonal specifically detects Septin-1 in Human samples. It is validated for Western Blot.Specifications
Septin-1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SEPT1 | |
Full length recombinant protein of human Septin-1 (NP_443070) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot | |
OTI1E1 | |
Western Blot | |
DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
1731.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction