Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin 1 Monoclonal Antibody (1F12)
GREENER_CHOICE

Catalog No. PIMA518728
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518728 Supplier Invitrogen™ Supplier No. MA518728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MDKEYVGFAA LPNQLHRKSV KKGFDFTLMV AGESGLGKST LINSLFLTNL YEDRQVPEAS ARLTQTLAIE RRGVEIEEGG VKVKLTLVDT PGFGDSVDCS DCWLPVVKFI EEQFEQYLRD ESGLNRKNIQ DSRVHCCLYF ISPFGRGLRP LDVAFLRAVH EKVNIIPVIG KADALMPQET QALKQKIRDQ LKEEEIHIYQ FPECDSDEDE DFKRQDAEMK ESIPFAVVGS CEVVRDGGNR PVRGRRYSWG TVEVENPHHC DFLNLRRMLV QTHLQDLKEV THDLLYEGYR ARRLQSLARP GARDRASRSK LSRQSATEIP LPMLPLADTE KLIREKDEEL RRMQEMLEKM QAQMQRSQAQ GEQSDAL.

Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin 1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1F12
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SEPTIN1
Gene Accession No. Q8WYJ6
Gene Alias 1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24
Gene Symbols SEPTIN1
Host Species Mouse
Immunogen SEPT1 (AAH12161, 1 a.a. ~ 367 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1731
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.