Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Septin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599715
Description
Antibody detects endogenous levels of total Septin 1.
Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.
Specifications
Septin 1 | |
Polyclonal | |
Unconjugated | |
SEPTIN1 | |
1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1731 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WYJ6 | |
SEPTIN1 | |
A synthesized peptide derived from human SEPT1(Accession Q8WYJ6), corresponding to amino acid residues E279-T329. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction