Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518577
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518577 Supplier Invitrogen™ Supplier No. PA518577
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, human, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Septin 1 (SEPT1) is a member of the septin family of GTPases. Members of this family are required for cytokinesis and the maintenance of cellular morphology. This gene encodes a protein that can form homo- and heterooligomeric filaments, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. Alternatively spliced transcript variants have been found but the full-length nature of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SEPTIN1
Gene Accession No. Q8WYJ6
Gene Alias 1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24
Gene Symbols SEPTIN1
Host Species Goat
Immunogen Synthetic peptide sequence (GARDRASRSKLSRQ) corresponding to the internal amino acids of 37135 (aa 306-320).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1731
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.