Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Septin-10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583886
Description
Immunogen sequence: MASSEVARHL LFQSHMATKT TCMSSQGSDD EQIKRENIRS LTMSGHVG.
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.
Specifications
Septin-10 | |
Polyclonal | |
Unconjugated | |
SEPT10 | |
4921515A04Rik; 9430099J10Rik; AA408298; AI874685; FLJ11619; SEPT10; septin 10; SEPTIN10; septin-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0V9 | |
SEPT10 | |
Recombinant protein corresponding to Human SEPT10. Recombinant protein control fragment (Product #RP-100558). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction