Learn More
Septin 10/SEPT10 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304916AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the septin family of cytoskeletal proteins with GTPase activity. This protein localizes to the cytoplasm and nucleus and displays GTP-binding and GTPase activity. Alternate splicing results in two transcript variants encoding different isoforms.Specifications
Septin 10/SEPT10 | |
Polyclonal | |
Unconjugated | |
SEPT10 | |
septin-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151011 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9P0V9 | |
SEPT10 | |
Between 404 and 454 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.