Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-11 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232159V
Description
Septin-11 Polyclonal antibody specifically detects Septin-11 in Human, Mouse samples. It is validated for Western BlotSpecifications
Septin-11 | |
Polyclonal | |
Western Blot | |
FLJ10849, septin 11, septin-11 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Septin 11 using the numbering given in entry NP_060713.1 (GeneID 55752). | |
0.1 mL | |
Cell Cycle and Replication | |
55752 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction