Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin-11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529435 Supplier Invitrogen™ Supplier No. PA529435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-11
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene SEPT11
Gene Accession No. Q9NVA2
Gene Alias 6230410I01Rik; AW548875; D5Ertd606e; SEPT11; Sept6; septin 11; septin 6; Septin11; septin-11
Gene Symbols SEPT11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Septin 11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55752
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.