Learn More
Septin 11 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304177AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking (Hanai et al., 2004 [PubMed 15196925]; Nagata et al., 2004 [PubMed 15485874]).Specifications
Septin 11 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8C1B7, Q9NVA2 | |
SEPT11 | |
Between 379 and 429 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SEPT11 | |
septin-11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
52398, 55752 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.