Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin-14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567282 Supplier Invitrogen™ Supplier No. PA567282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPTDEETAAQ ANSSVSGLL Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.

SEPT14 is a member of the highly conserved septin family of GTP-binding cytoskeletal proteins implicated in membrane transport, apoptosis, cell polarity, cell cycle regulation, cytokinesis, and other cellular functions.[supplied by OMIM, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPT14
Gene Accession No. Q6ZU15
Gene Alias 14-Sep; 1700016K13Rik; Sept14; septin 14; SEPTIN14; septin-14
Gene Symbols SEPT14
Host Species Rabbit
Immunogen Recombinant Human Septin-14. Recombinant protein control fragment (Product #RP-107920).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.