Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101718 Supplier Invitrogen™ Supplier No. PA5101718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SEPT3.

This gene belongs to the septin family of GTPases. Members of this family are required for cytokinesis. Expression is upregulated by retinoic acid in a human teratocarcinoma cell line. The specific function of this gene has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin 3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SEPT3
Gene Accession No. Q9UH03, Q9WU34, Q9Z1S5
Gene Alias 3110018K01Rik; 3-Sep; AV154067; B530002E20Rik; bK250D10.3; CTA-250D10.3; G-septin; G-septin alpha; MGC133218; neuronal-specific septin 3; neuronal-specific septin-3; OTTHUMP00000028742; P40; Sep3; SEPT3; septin 3; Septin3
Gene Symbols SEPT3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SEPT3(Accession Q9UH03), corresponding to amino acid residues N135-L185.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24050, 55964, 56003
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.