Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312342
This item is not returnable.
View return policy
Description
Septin-5 Polyclonal antibody specifically detects Septin-5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Septin-5 | |
Polyclonal | |
Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:50-1:150, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50-1:150 | |
CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
Synthetic peptide taken within amino acid region 300-350 on SEPT5 protein. | |
100 μg | |
Cell Cycle and Replication | |
5413 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction