Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-5 Antibody (SP18), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281058UV
Description
Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Septin-5 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
SEPTIN5 | |
Immunoprecipitate from human brain. | |
0.1 mL | |
Cell Cycle and Replication | |
5413 | |
Human, Rat, Rabbit | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
SP18 | |
Western Blot, ELISA, Immunohistochemistry | |
CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2. | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction