Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-5, Rabbit anti-Human,Rat,Rabbit, Clone: SP18, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Septin-5 |
---|---|
Clone | SP18 |
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA, Immunohistochemistry |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Septin-5 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS | |
5413 | |
Immunoprecipitate from human brain. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
SP18 | |
Western Blot, ELISA, Immunohistochemistry | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Rabbit | |
CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
SEPTIN5 | |
IgG κ | |
Protein A purified | |
This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title