Learn More
Septin 7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304212AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Orangutan and Chimpanzee
This gene encodes a protein that is highly similar to the CDC10 protein of Saccharomyces cerevisiae. The protein also shares similarity with Diff 6 of Drosophila and with H5 of mouse. Each of these similar proteins, including the yeast CDC10, contains a GTP-binding motif. The yeast CDC10 protein is a structural component of the 10 nm filament which lies inside the cytoplasmic membrane and is essential for cytokinesis. Although the exact function of this gene has not yet been determined, its high similarity to yeast CDC10 and the high conservative nature of eukaryotic cell cycle machinery suggest a similar role to that of its yeast counterpart. Alternative splicing results in two transcript variants encoding different isoforms.Specifications
Septin 7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O55131, Q16181 | |
SEPT7 | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SEPT7 | |
SEPT7; septin 7; CDC10; CDC3; Nbla02942; SEPT7A; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 cell division cycle 10 homolog; cell division cycle 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
235072, 989 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.