Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-7 Rabbit anti-Human, Mouse, Rat, Clone: S09-6A1, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Septin-7 |
---|---|
Clone | S09-6A1 |
Dilution | Western Blot, ELISA, Immunohistochemistry-Paraffin 20 μg/mL, Immunofluorescence 20 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Monoclonal |
Description
Septin-7 Monoclonal antibody specifically detects Septin-7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
Septin-7 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin 20 μg/mL, Immunofluorescence 20 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CDC10 cell division cycle 10 homolog, CDC10 cell division cycle 10 homolog (S. cerevisiae), CDC10 protein homolog, CDC10S. cerevisiae, homolog), Nbla02942, septin 7, septin-7 | |
Recombinant protein of human Septin-7 (Uniprot#: Q16181) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S09-6A1 | |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
989 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title