Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin-8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583339 Supplier Invitrogen™ Supplier No. PA583339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAFNRRKAAV EALQSQALHA TSQQPLRKDK DKKNRSDIGA HQPGMSLSSS KVMMTKASVE PLNCSSWWPA IQCCSCLVRD ATWREGFL Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 91%.

SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 [PubMed 12909369]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SEPTIN8
Gene Accession No. Q92599
Gene Alias 2-Sep; AW046166; EMBL:AAI58797.1, ECO:0000312; KIAA0202; mKIAA0202; RGD:1308449}; SEP2; Sepl; Sept8; sept8 {ECO:0000312; septin 8; Septin8; septin-8; septin-8 {ECO:0000250; UniProtKB:Q8CHH9}
Gene Symbols SEPTIN8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SEPT8. Recombinant protein control fragment (Product #RP-104667).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23176
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.