Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin-8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590178 Supplier Invitrogen™ Supplier No. PA590178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKVKETELEL KEKERELHEK FEHLKRVHQE EKRKVEEKRR ELEEETNAFN RRKAAVEALQ SQALHATSQQ PLRKDKDKKN; Positive Samples: U-251MG, HeLa, LO2, Mouse heart, Mouse kidney, Rat brain; Cellular Location: Cytoplasm, cytoskeleton.

SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 [PubMed 12909369]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-8
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SEPTIN8
Gene Accession No. B0BNF1, Q8CHH9, Q92599
Gene Alias 2-Sep; AW046166; EMBL:AAI58797.1, ECO:0000312; KIAA0202; mKIAA0202; RGD:1308449}; SEP2; Sepl; Sept8; sept8 {ECO:0000312; septin 8; Septin8; septin-8; septin-8 {ECO:0000250; UniProtKB:Q8CHH9}
Gene Symbols SEPTIN8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 348-427 of human Septin 8 (NP_0012877281).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20362, 23176, 303135
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.