Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Septin-8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599716 Supplier Invitrogen™ Supplier No. PA599716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Septin 8.

SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 [PubMed 12909369]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Septin-8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SEPTIN8
Gene Accession No. Q92599
Gene Alias 2-Sep; AW046166; EMBL:AAI58797.1, ECO:0000312; KIAA0202; mKIAA0202; RGD:1308449}; SEP2; Sepl; Sept8; sept8 {ECO:0000312; septin 8; Septin8; septin-8; septin-8 {ECO:0000250; UniProtKB:Q8CHH9}
Gene Symbols SEPTIN8
Host Species Rabbit
Immunogen A synthesized peptide derived from human SEPT8(Accession Q92599), corresponding to amino acid residues R289-R339.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23176
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.