Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Sequenase Version 2.0 DNA Polymerase

Catalog No. p-7138029
Encompass_Preferred
Click to view available options
Quantity:
1000 U
200 U

Sequenase™ Version 2.0 DNA Polymerase is a genetically engineered form of T7 DNA polymerase. Unlike the wild-type enzyme it has virtually no 3'→5' exonuclease activity.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 13 units/μL
Description Sequenase Version 2.0 DNA Polymerase, 200 Units, 13 units/μL Concentration, Genetically-Engineered form of T7 DNA Polymerase, 7.5 pH, 37°C Optimum Temperature
For Use With (Application) Sequencing
Polymerase Genetically-Engineered form of T7 DNA Polymerase
Product Type Sequenase Version 2.0 DNA Polymerase
Quantity 200 U
Purity 0.95
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.